KIMBLE® Outer Joint Hose Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
182910-2440 276110-1420
KT182910-2440EA 122.63 CAD
KT182910-2440 KT276110-1420
KIMBLE® Outer Joint Hose Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Ideal for use as a vent or for the introduction of pressure/vacuum.
  • [ST] joint at one end
  • Vertical hose connection at other end
  • Manufactured from borosilicate glass
Adapter features a Standard Taper joint at one end and vertical hose connection at the other.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.