KIMBLE® Offset BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
179710-2119 179710-2124
KT179710-2119CS 109.94 CAD
KT179710-2119 KT179710-2124 KT179710-2114
KIMBLE® Offset BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
A unique glass adapter offset and angled approximately 10° for use with multi-neck flasks.
  • Offset and angled approximately 10°
  • Vacuum-tight seal
  • Open-top compression cap and FKM O-Ring included
Thermometers or bleed tubes are angled into the bottom of a flask
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
A vacuum-tight seal is created with the open-top compression cap and FKM O-Ring to allow adjustable immersion of thermometers. Cap is suitable for use to 150°C (302°F). Supplied complete with one size 010 FKM O-Ring and one cap.