Biotube™ Cluster Tube System, Simport
Supplier: Simport Scientific
T100-26 T100-28 T100-40 T100-2B T100-4B T100-4G T100-4R T100-20 T100-25 T100-35 T100-2G T100-2R T100-2Y T100-3B T100-3G T100-3R T100-3Y T100-4Y T100-50B T100-50G T100-50R T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-1B T100-1G T100-1R T100-1Y T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-50B T100-50G T100-50R T100-20LST
CA60870-021CS 294.56 CAD
CA60870-021 CA60870-026 CA60870-028 CA60872-504 CA60872-512 CA60872-513 CA60872-514 CA60872-489 CA60872-490 CA60872-494 CA60872-505 CA60872-506 CA60872-507 CA60872-508 CA60872-509 CA60872-510 CA60872-511 CA60872-515 CA60872-520 CA60872-521 CA60872-522 CA60872-523 CA60872-524 CA60872-525 CA60872-526 CA60872-527 CA60872-684 CA60872-686 CA60872-688 CA60872-690 10199-456 10199-544 10199-546 10199-548 10199-550 10199-558 10199-630 10199-632 10199-640
Biotube™ Cluster Tube System, Simport
Tubes Microtubes
The Biotube™ Cluster Tube System is a polypropylene tube and rack system with tubes held in a standard 8×12 configuration.
  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Ideal for storing, freezing and transporting reagents and specimens
  • Racks and tubes can be used over a wide temperature range, −80 to 121 °C
Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as a self-standing support. It can also be placed in a refrigerator or freezer for improved air circulation around the tubes, or in water baths to allow controlled warming of the tubes contents.