FTA® Cards, Qiagen
Supplier: Qiagen
WB120205 WB120210 WB120065 WB120056 WB120208 WB120206 WB120056 WB120211 WB120208 WB120355 WB120306 WB120305 WB120311 WB120356 WB120310
14222-800PK 2414.11 CAD
14222-800 14222-808 80077-200 CA14222-806 CA14222-812 14222-802 14222-804 14222-806 14222-810 14222-812 CA11013-648 CA11013-650 CA11013-652 CA11013-656 CA11013-664 CA11014-012 10035-570 10035-868 10035-870 10035-872
FTA® Cards, Qiagen
Nucleic Acid Storage Systems
For the collection, transport, archiving and isolation of nucleic acids at room temperature.
  • Nucleic acids are captured, immobilized, and stabilized for storage, all in one easy step
  • Captured nucleic acid is ready for downstream applications in less than 30 minutes
  • Different card options to support manual or automated processing for fast and efficient sampling
  • Nucleic acids collected on FTA Cards remain stable for years at room temperature
  • Indicating FTA Cards change color upon sample application to facilitate handling of colorless samples