Non-Indicating FTA® Elute Micro Cards, Qiagen
Supplier: Qiagen
WB120401 WB120411 WB120411 WB120410 WB120401 WB120412
CA89012-926PK 541.46 CAD
CA89012-926 CA95059-622 95059-622 89012-928 89012-926 95059-624
Non-Indicating FTA® Elute Micro Cards, Qiagen
Nucleic Acid Storage Systems
FTA® Elute Cards utilise FTA® technology that simplifies the handling and processing of nucleic acids. DNA is eluted in an easy step, providing DNA in solution ready for amplification. Available as non-indicating cards for use with colored samples, such as blood.
  • Samples can be collected, shipped, and preserved at room temperature, eliminating the high costs associated with shipping samples on ice and with laboratory freezer storage
  • Sample processing time to isolate DNA of 15 to 30 minutes, thanks to a simple hot water elution procedure eliminating the
More Product Information