BRAND PCR Plates for Roche® LightCycler® 480, BrandTech®
Supplier: Brandtech
781358 781364 781365
89135-448PK 2448.51 CAD
89135-448 89400-704 89400-706
BRAND PCR Plates for Roche® LightCycler® 480, BrandTech®
Microplates PCR Plates
These white PCR plates provide high signal to noise for real-time/qPCR.
  • Uniformally coloured with titanium dioxide (TiO₂)
  • Smooth surfaces to optimise reflection of the fluorescence signal
  • Extra-thin wall thickness to provide optimal thermal transfer and short cycle times
  • 96-well plates have black alphanumeric code, no additional markings on the cut corner
  • Manufactured under modern cleanroom conditions according to ISO 14 644-1 class 5 to 8
  • DNase-, DNA- and RNase-free
Available with 96-well low profile and 384-well formats. Both plates are compatibleMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable