KimCote® Cylinders with Reverse Graduations, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
KC20028W-100 KC20028W-250 KC20028W-500 KC20028W-1000 KC20028W-2000
89097-382CS 192.86 CAD
89097-382 89097-384 89097-386 89097-388 89097-390
KimCote® Cylinders with Reverse Graduations, Class A, Kimble Chase
Cylinders Measuring Cylinders
KimCote® safety coating reduces the hazards of shattered glass
Designed from ASTM Specification E1272, Style I, Class A requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Each cylinder is marked with “TD” which indicates the cylinder is calibrated to deliver, and a white, reverse, single metric scale. The hexagonal base is flat ground for stability and the SAFE-GARD® bumper is supplied.
Order Now
SPECIFICATIONS
Form Tall
Environmentally Preferable