VWR® Ice Pans
Supplier: VWR International
10146-202 10146-204 10146-206 10146-208 10146-210 10146-212 10146-214 10146-216 10146-218 10146-220 10146-222 10146-224 10146-226 10146-228 10146-230 10146-232 10146-234 10146-236 10146-278 10146-280 10146-282 10146-294 10146-296 10146-298
10146-202EA 91.56 CAD
10146-202 10146-204 10146-206 10146-208 10146-210 10146-212 10146-214 10146-216 10146-218 10146-220 10146-222 10146-224 10146-226 10146-228 10146-230 10146-232 10146-234 10146-236 10146-278 10146-280 10146-282 10146-294 10146-296 10146-298
VWR® Ice Pans
Ice Containers
VWR® Ice Pans are manufactured from polyurethane foam to ensure good insulating properties.
  • Polyurethane foam construction
  • pouring lip for easy draining
  • Lids can be stacked beneath pans
Ice Pans are suitable for use with Dry-Ice with Isopropyl Alcohol, Water-Ice with salt slurry, or liquid nitrogen. The operating temperature range is -76 to 70°C (-104 to 158°F). All pans feature a pouring lip for easy draining. VWR® Ice Pans are available with or without lids. The lids, which are unbreakable in normal use, help to reduce evaporation levels. Lids can also be stacked below the pans for convenient, space-saving storage.