KIMBLE® Straight Connecting Adapters, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
166000-7524 166100-3024
KT166000-7524EA 168.85 CAD
KT166000-7524 KT166100-3024 KT166000-7529
KIMBLE® Straight Connecting Adapters, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Straight Connecting Adapters expand the length of an apparatus, and are also useful as an air condenser.
  • 155mm and 160mm sizes have an outer [ST] joint at one end and inner [ST] joint on the other end
  • 110mm size has inner [ST] joints at both ends
  • Manufactured from borosilicate glass
The 155mm and 160mm adapters feature outer Standard Taper joints at one end and inner Standard Taper joints on the other end
Adapters are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class AMore Product Information