KIMBLE® Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747125-1314 747125-2019
KT747125-1314EA 106.72 CAD
KT747125-1314 KT747125-1319 KT747125-1324 KT747125-2014 KT747125-2019 KT747125-2024
KIMBLE® Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Adapters allow interfacing between outer Standard Taper jointed glassware with threaded microscale components.
  • Threaded on one end
  • With [ST] inner joint
  • Manufactured from borosilicate glass
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.