VERSA 110 PCR Set-up Workstation, Aurora Biomed
Supplier: Aurora Biomed
VER-110-PCR
CA11023-070EA 60751.72 CAD
CA11023-070
VERSA 110 PCR Set-up Workstation, Aurora Biomed
Fume Hoods and Enclosures PCR Workstation Enclosures
Automation of PCR setup allows labs to increase throughput, reduce variation, improve reproducibility and provide valuable walk-away time for lab staff to focus on other important tasks
Featured include a complete walk-away solution that frees up time for lab staff, speedier workflow to get results sooner, open system that allows leverage of inexpensive 3rd party reagents and high precision that reduces sample variability and increases reproducibility. Additional benefits include low dead volume that reduces costs associated with reagent waste, HEPA enclosure that protects sensitive samples from outside contamination, decreased contact with lab staff reducing the chance for iatrogenic infection and flexibility to perform other associated tasks such as normalization, plate reformatting and more.
Aurora’s VERSA series of automated PCR setup workstations utilize highly accurate air displacement technology to provide the accuracy and precision required by modern labs. With modular components such as cooling and heating plates, shakers, reagent cooling blocks, and others, the VERSA liquid handling workstations are a highly flexible platform to assist in a variety of laboratory workflows.
Certifications: CSA Certified
Ordering information: Computer and accessories sold separately.