TRANSFORM Microwave Digestion Systems, Aurora Biomed

Supplier: Aurora Biomed
0680-MW-1680 0800-MW-1800
CA11023-062EA 44695.99 CAD
CA11023-062 CA11023-064
TRANSFORM Microwave Digestion Systems, Aurora Biomed
Digestion Systems Mineralisation Workstations
Aurora’s TRANSFORM series of microwave digestion systems offer intelligent, rapid digestion of a variety of sample types with some of the most advanced safety features available on the market

Features include round body made of PFA-coated stainless steel for highest blast resistance, evenly distributed microwave field throughout the chamber, uniform heat for each sample vessel, top-loading automatic cover, direct sensing and control of sample temperature in closed vessels and reproducible digestion. Additional features include full compliance with EPA methods, on-line directory for EPAMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Electrical 220V / 12A 220V / 12A
Power 1200 (W) 1200 (W)
Weight 30 kg (66 lbs.) 50 kg (110 lbs.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR