TRACE AI1200 Atomic Absorption Spectrometers, Aurora Biomed
Supplier: Aurora Biomed
1200-S-F 1200-S-GF 1200-S-F-GF
CA11023-052EA 49507.5 CAD
CA11023-052 CA11023-054 CA11023-056
TRACE AI1200 Atomic Absorption Spectrometers, Aurora Biomed
Spectrophotometers and Photometers Flame Photometers
The TRACE series of atomic absorption spectrometers deliver ultra-trace level detection of over 70 different elements making them a valuable instrument in almost any laboratory environment
Features include automatic Ar-H2 flame ignition, automatic switching between flame and graphite furnace atomization, transversely heated GF tube with high heating rate, inert Teflon nebulizer chamber, universal XYZ autosampler, auto-aligning coded lamp turret and on-line dilution.
The TRACE series of atomic absorption spectrometers are used in agricultural, clinical, environmental, forensics,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable