PTFE Plug-Style Stopcocks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
41502T-2 821001-0002 821001-0004 821001-0006
59180-140CS 241.43 CAD
59180-140 KT821001-0002 KT821001-0004 KT821001-0006
PTFE Plug-Style Stopcocks, DWK Life Sciences
Stopcocks
PTFE stopcock plug has a 1:5 taper and is chemically-resistant and self-lubricating.