Air Rail, Production Basics
Supplier: Production Basics
PB-8330 PB-8336 PB-8333
76239-066EA 366.8 CAD
76239-066 76239-068 76240-294
Air Rail, Production Basics
Furniture Casework
Features two quick-connectors and one 0.9525 cm (0.375" or ³/₈") poly-tube inlet port for compressed air.
  • Provides reserve air capacity at workstation
  • Can be mounted on the uprights or overhead arms
For use with on any C-leg lab bench. Add uprights to mount accessories on RTW lab bench, easy lift dual-leg, PedSys lab bench, basic organizer or easy lift 4-post lab bench.
Ordering information: Standard accessory color is black. Other standard frame color (light neutral/almond, blue, white, or grey) available by request. Accessory is ready to mount onMore Product Information