Easy-Lift Dual Leg Workstation, ESD, Production Basics
Supplier: Production Basics
PB-4300 PB-4302
76243-136EA 9095.29 CAD
76243-136 76246-320
Easy-Lift Dual Leg Workstation, ESD, Production Basics
Furniture Tables and Workbenches
Ideal for two or three-shift operations requiring sitting and standing laboratory tasks in a sensitive environment.
  • Finished with rounded corners and protective T-mold edge
  • Store up to four height settings
  • Measures within static dissipative range of 10⁶ to 10⁹
Vertical movement through electric hydraulic system connects to electrical outlet for immediate productivity. Effortless height 76.2 to 106.7 cm (30 to 42") adjustment with a push button travels the full range within 20 seconds.
Ordering information: Frame is Black epoxy powder coat with extruded aluminum legs. Other standard frame color (Light Neutral/Almond, Blue, White, or Grey) available by request. Specify a Laminate color otherwise will be default Grey ESD Laminate. Worksurface available in White, Grey and Beige. Workstation ships assembled. Accessories are available and sold separately; uprights (models 8539, 8540, 8532 or 8533) are needed for above surface accessories.
Delivery information: Includes (1) easy-lift frame, (1) ESD worksurface, (1) ground bolt and wire, (1) electric hydraulic module, (4) leveling feet.