Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]

Supplier: Tonbo Biosciences
70-0281-U025 70-0281-U100 70-0281-U500
10051-674EA 36.34 CAD
10051-674 10051-676 10051-678
Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]
Antibodies
The 37.51 antibody reacts with mouse CD28, a 45 kDa glycoprotein which acts as a co-stimulatory receptor in support of the T cell receptor (TCR). CD28 exists as a homodimer with specificity for two known ligands, known as B7-1 (CD80) and B7-2 (CD86), expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. Other members of the CD28 family of co-stimulatory receptors include CTLA-4 (CD152), PD-1 (CD279), ICOS and BTLA.

The 37.51 may be used as a phenotypic marker for CD28, which is expressed on all CD4+ T cells and CD8+ T cells, and on NK cells in mouse. In addition, the 37.51 antibody is widely used to activate the CD28 receptor in vitro and in vivo. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation: Unconjugated
Epitope:
Host: Golden Syrian Hamster
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR