Anti-FASLG Mouse Antibody (APC (Allophycocyanin)) [clone: NOK-1]

Supplier: Tonbo Biosciences
20-9919-T025 20-9919-T100
10141-006EA 118.5 CAD
10141-006 10141-008
Anti-FASLG Mouse Antibody (APC (Allophycocyanin)) [clone: NOK-1]
Antibodies
The NOK-1 antibody reacts with human CD178 (Fas ligand) in both membrane bound and soluble forms. Fas ligand is a 40 kDa transmembrane glycoprotein, a member of the TNF family, and is expressed by activated T and NK cells, neutrophils, and monocytes. Interactions between CD178 (Fas ligand) and CD95 (Fas) induce a program of apoptosis and play a key role in immune regulation and homeostasis. The extracellular domain of human CD178 can be cleaved from the surface by matrix metalloproteinases (MMPs) resulting in a 26 kDa soluble protein.

Type: Primary
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR