Anti-CD3E Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Hit3a]

Supplier: Tonbo Biosciences
35-0039-T025 35-0039-T100
10050-816EA 37.92 CAD
10050-816 10050-818
Anti-CD3E Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Hit3a]
Antibodies
The Hit3a antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing cell proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.

The Hit3a antibody is a widely used phenotypic marker for human T cells. In addition, binding/cross-linking of Hit3a antibody to CD3e can induce cell activation. The antibody has also been demonstrated to be cross-reactive with Chimpanzee CD3. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: Hit3a
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR