Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]

Supplier: Tonbo Biosciences
20-1172-U025 20-1172-U100
10050-566EA 85.32 CAD
10050-566 10050-568
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]
Antibodies
The ACK2 antibody is specific for CD117, also called c-Kit, a 145 kDa cytokine receptor important in the development of hematopoietic stem cells, in oogenesis, and for functional activity of immune cells such as NK and mast cells. c-Kit binds to a ligand known as stem cell factor (SCF), or alternatively as mast cell growth factor. Ligand binding promotes the activation (dimerization) and subsequent tyrosine kinase activity of the c-Kit receptor and triggers key survival, expansion and maturation signals during hematopoietic progenitor cell development. Conversely, shedding of extracellular domain of c-Kit receptor is reported to induce inactivation or apoptosis within these cells. The survival signaling activity of c-Kit confers a proto-oncogenic attribute to the receptor, as overexpression or mutations in this protein are associated with tumor development.

The ACK2 antibody is widely utilized as a marker to identify hematopoietic progenitors, and to neutralize receptor-ligand binding in vitro and in vivo (use format appropriate for functional assays). In addition, the antibody is reported to be cross-reactive with rat c-Kit and is extensively published for use in this species.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: ACK2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR