Anti-SELL Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEL-14]

Supplier: Tonbo Biosciences
50-0621-U025 50-0621-U100
10051-270EA 71.1 CAD
10051-270 10051-272
Anti-SELL Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEL-14]
Antibodies
The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte “rolling” on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol SELL
Environmentally Preferable
Antigen name Selectin L
Antigen synonyms LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone MEL-14
Reactivity Mouse
Host Rat
Gene ID 20343
Isotype IgG2a
Flow cytometry Yes
Neutralisation Yes
Storage temperature Store undiluted at 2-8°C, protect from light, do not freeze.
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR