Anti-PTPRC Rat Monoclonal Antibody (violetFluor® 450) [clone: 104]

Supplier: Tonbo Biosciences
75-0454-U025 75-0454-U100
10050-546EA 97.96 CAD
10050-546 10050-548
Anti-PTPRC Rat Monoclonal Antibody (violetFluor® 450) [clone: 104]
Antibodies
The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors. 

The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 104
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR