Corning® Human Recombinant Laminin-521
Supplier: Corning
rLaminin-521 (human) is an animal-component free recombinant heterotrimer composed of α5, β2 and γ1 chains expressed in a mammalian cell culture system. rLaminin-521 supports long-term self-renewal of human pluripotent stem cells (hPSC) -hESC & ihPSC and Neural Stem Cells(NSCs) in defined media environment.
- Expressed in mammalian cell culture system
- Supports long- term self-renewal of human pluripotent stem cells (hPSC)
- Provides ROCK inhibitor independent single cell expansion of PSCs, which improves hPSC culture ease and efficiency
- Easy to self-coat
- Compatible with multiple hPSC culture media and reagents
- Supports undifferentiated expansion of hPSC
- Suitable for xeno-free, defined culture environment
Sterility: Non-pyrogenic
Caution: Unless otherwise specified, all products are for research use only. Not intended for use in diagnostic or therapeutic procedures. No claims regarding the performance of these products for clinical or diagnostic applications.