Anti-LY6C1 Rat Antibody (redFluor® 710) [clone: RB6-8C5]
Supplier: Tonbo Biosciences
80-5931-U025 80-5931-U100
10141-122EA 94.8 CAD
10141-122 10141-124
Anti-LY6C1 Rat Antibody (redFluor® 710) [clone: RB6-8C5]
Antibodies
The RB6-8C5 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.
The RB6-8C5 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: The RB6-8More Product Information