Ocean Pacific® Blue Lite™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
OPXS-BL2 OPS-BL2 OPM-BL2 OPL-BL2 OPXL-BL2
CA10066-420CS 416.99 CAD
CA10066-420 CA10066-422 CA10066-424 CA10066-426 CA10066-428
Ocean Pacific® Blue Lite™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Gloves
Lightweight, powder-free nitrile glove that fits and feels like latex.
  • Lightweight design provides exceptional comfort through the palm and thumb
  • Additional thickness at fingertips ensures durability in the most crucial areas
  • Internal surface is polymer coated for improved donning and biocompatibility
  • Textured fingertips and bonded polymer exterior surface ensure a secure grip, even when wet
Ideal when protection against mechanical or biological agents is required. Ultra-light for optimal dexterity and tactility. Textured fingertips ensure a secureMore Product Information