Aqua Blue® Nitrile Gloves, Powder Free, Medicom
Supplier: Avantor
CA10144-576CS 521.05 CAD
CA10144-576 CA10144-578 CA10144-580 CA10144-582 CA10144-642
Aqua Blue® Nitrile Gloves, Powder Free, Medicom
The rugged durability you have come to rely upon is now available acceslerator-free, in the new 4.4 mil thickness.
  • Unique nitrile compound sheds water better for a more secure grip, even in wet environments
  • Tested to be safe for use with chemotherapy drugs
Created using the latest in powder-free, accelerator-free nitrile, it has fewer chemicals and virtually eliminates the risk of Type 1 latex, and Type IV chemical allergic reactions and contact dermatitis.
Aqua Blue is now thinner, more flexible and form-fitting to reduce wear fatigue without compromising hand protection from mechanical and chemical hazards.