Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Monoclonal Antibody [clone: M5/114.15.2] (APC)
Supplier: Tonbo Biosciences
20-5321-U025 20-5321-U100
10050-618EA 71.1 CAD
10050-618 10050-620
Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Monoclonal Antibody [clone: M5/114.15.2] (APC)
Antibodies
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.
The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also beMore Product Information