KimCote® Class A Heavy Duty Wide Mouth Volumetric Flasks, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
KC92812G-100 KC92812G-250 KC92812G-500 KC92812G-1000 KC92812G-2000
89217-702CS 1241.14 CAD
89217-702 89217-704 89217-706 89217-708 89217-710
KimCote® Class A Heavy Duty Wide Mouth Volumetric Flasks, Kimble Chase
Flasks Volumetric Flasks
Wide mouth volumetric flasks feature an autoclavable KimCote® safety coating that reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
KimCote® volumetric flasks are graduated to Class A volumetric tolerances and are designed from ASTM Specification E288. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Superior strength, durability and heavy uniform walls ensure safety in the lab with neck dimensions that will easily accommodate pipets. Flat interior bottoms are ideal for stirring with standard stir bars. Calibrated to contain with Standard Taper glass stoppers.