Anti-FCER1A Armenian Hamster Monoclonal Antibody

Supplier: Tonbo Biosciences
40-5898-U100 40-5898-U500
10051-114EA 188.02 CAD
10051-114 10051-116
Anti-FCER1A Armenian Hamster Monoclonal Antibody
Antibodies
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.

The MAR-1 antibody is widely used both in flow cytometry and for depletion of cells in vitro / in vivo. Please choose the appropriate format for each application.

Type: Primary
Antigen: Fc epsilon Receptor I alpha (FceR1)
Clonality:
Clone:
Conjugation: In Vivo Ready™
Epitope:
Host: Hamster (Armenian)
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR