Anti-LPAM-1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: DATK32]

Supplier: Tonbo Biosciences
20-5887-U025 20-5887-U100
10050-630EA 105.86 CAD
10050-630 10050-632
Anti-LPAM-1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: DATK32]
Antibodies
The DATK32 antibody is specific for a combinatorial epitope of the mouse integrin alpha 4/beta 7 (LPAM-1) heterodimer. LPAM-1 is expressed on most mature lymphocytes and sub-populations of thymocytes and bone marrow cells. It is involved in lymphocyte adhesion and migration and is a receptor for several extracellular matrix proteins including VCAM-1, fibronectin, and MAdCAM-1.

DATK32 is reported to induce LPAM-1 dependent lymphocyte aggregation and block adhesion to VCAM-1, fibronectin, and MAdCAM-1.

Type: Primary
Antigen: LPAM-1
Clonality: Monoclonal
Clone: DATK32
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR