Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F1]
Supplier: Tonbo Biosciences
50-5893-U025 50-5893-U100
10051-354EA 99.54 CAD
10051-354 10051-356
Anti-KLRG1 Golden Syrian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F1]
Antibodies
The 2F1 antibody reacts with mouse KLRG1 (Killer cell Lectin-like Receptor G1). This 30-38 kDa homodimeric receptor may be expressed by activated, mature NK cells and by effector/memory T cells, with potentially different roles in each cell type. KLRG1 can regulate, in an inhibitory fashion, the development and effector functions of NK cells, and is often cited as a senescence or terminal differentiation marker for T cells. Ligands for KLRG1 include members of the cadherin family of adhesion molecules, specifically N-Cadherin, E-Cadherin, and R-Cadherin. These interactions may induceMore Product Information