Sterile Rabbit Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D509-04-0050
D509-06-0050
D509-08-0050
CAD509-04-0050EA
1158
CAD
CAD509-04-0050
CAD509-06-0050
CAD509-08-0050
Sterile Rabbit Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.