Sterile Swine Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D517-04-0050 D517-06-0050 D517-08-0050
CAD517-04-0050EA 923 CAD
CAD517-04-0050 CAD517-06-0050 CAD517-08-0050
Sterile Swine Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.