Sterile Sheep Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D511-04-0050
D511-06-0050
D511-08-0050
CAD511-04-0050EA
471.99
CAD
CAD511-04-0050
CAD511-06-0050
CAD511-08-0050
Sterile Sheep Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.