Sterile Rat Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D510-04-0050
D510-05-0050
D510-06-0050
D510-08-0050
CAD510-04-0050EA
855
CAD
CAD510-04-0050
CAD510-05-0050
CAD510-06-0050
CAD510-08-0050
Sterile Rat Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.