Sterile Guinea Pig Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D505-04-0050
D505-06-0050
D505-08-0050
CAD505-04-0050EA
1419.99
CAD
CAD505-04-0050
CAD505-06-0050
CAD505-08-0050
Sterile Guinea Pig Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.