Sterile Goat Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D504-04-0050 D504-06-0050
CAD504-04-0050EA 471.99 CAD
CAD504-04-0050 CAD504-06-0050
Sterile Goat Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.