Sterile Calf Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D500-06-0050
D500-06-0100
CAD500-06-0050EA
356.01
CAD
CAD500-06-0050
CAD500-06-0100
Sterile Calf Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.