20-Place Cardboard Slide Trays
Supplier: Heathrow Scientific
HS9902 HS120196 HS120197
490012-384CS 2486.25 CAD
490012-384 10126-324 10126-326
20-Place Cardboard Slide Trays
Slide Holders
Cardboard slide folders offer a convenient solution for safely working, transporting and short-term storing of slides.
  • Compartment holder is made from a double layer of densely pressed cardboard
  • Cover is single layer cardboard
  • Front protection flaps are attached with durable tape
  • Recessed slide compartments and inside covers for specimen protection
  • Slides cannot be accessed when cover is closed reducing contamination
Index on the front cover corresponds with the number grid inside for identification. Cardboard slide trays hold standard slides 76×26 mm (3×1").
Order Now
SPECIFICATIONS
Environmentally Preferable