Combo Brooms, Soft/Stiff, Remco Products
Supplier: Remco Products
31742 31743 31744 31745 31746 31749
76007-148EA 101.8 CAD
76007-148 76007-150 76007-152 76007-154 76007-156 76007-162
Combo Brooms, Soft/Stiff, Remco Products
Brooms
This fully color-coded sweeping broom has two types of bristles to pick up every particle
The stiff bristles loosen stubborn dirt, and the soft bristles sweep away loose particles. Block material is polypropylene; bristle material is polyester.
Certifications: FDA Compliant Raw Material.