Lobby Brooms, Remco Products
Supplier: Remco Products
31044 31045 31046 31042 31043
76007-140EA 42.34 CAD
76007-140 76007-142 76007-144 76007-320 76007-322
Lobby Brooms, Remco Products
Brooms
This fully color-coded, soft-bristled lobby broom is great for dry areas and sweeping in between narrow spaces in production lines
Block material is polypropylene; bristle material is polyester.
Certifications: FDA Compliant Raw Material.
Order Now
SPECIFICATIONS
Environmentally Preferable