Heratherm Advanced Protocol Microbiological Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
51028066 51028067 51028068
10124-574EA 6636 CAD
10124-574 10124-592 10124-604
Heratherm Advanced Protocol Microbiological Incubators, Thermo Scientific
Incubators Standard Incubators
Successful incubation is dependent upon environmental conditions
Heratherm Advanced Protocol Microbiological Incubators feature 3 bench top sizes, a wide temperature range of ambient +5°C to 105°C, microprocessor control, rounded chamber corner for easy cleaning, automatic over-temperature alarm, internal outlet and access port, corrosion resistant AISI 304 Stainless Steel interior, RS232 interface, internal glass door, easy calibration routine, timer (weekly / daily / real time) and an interior outlet. Each unit has a flexible shelving system with no tools required and includes two perforated stainless steel shelves standard.
Dual convection technology allows the user to switch from gravity or forced air convection within the same unit. When using forced air, the fan speed is adjustable from 0 -100% in 20% increments. Excellent temperature uniformity as good as ± 0.2 °C and outstanding temperature stability at ± 0.1 °C.
As such, you need to trust that your valuable samples are maintained in a safe and efficient manner. Heratherm microbiological incubators are designed to provide long-term performance with optimal conditions. They are backed by a two-year warranty, meaning confidence for your investment. Heratherm microbiological incubators are designed with a very small footprint ratio compared to the interior volume. In addition they are stackable without the need of any tools or stacking devices.
Certifications: cCSAus