Sedimat® 15 Plus Automated Sedrate System, Polymedco
Supplier: Polymedco
Sedimat®
SJ-1215 PLUS SJ-1215 PLUS
CA89423-716EA 12861.72 CAD
CA89423-716 89423-716
Sedimat® 15 Plus Automated Sedrate System, Polymedco
Clinical Diagnostic Supplies Clinical Instruments
The Sedimat® 15 Plus is an automated sedrate system that provides accurate ESR results in only 15 minutes with optional bar code reader and LIS interface capability
Offering random access, great value, and low cost, the system allows faster throughput (process 32 patient results per hour) in a small footprint. The 8-position reader provides computer interfacing to transfer data.
Fast, in-office results which can test one to eight specimens simultaneously
Ordering information: Accessories not included.