ACTERO™ Other Application Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC. CA
FCM-150 FCM-169 FCM-160 FCM-157 FCM-069 FCM-165 FCM-072 FCM-171 FCM-172 FCM-156 FCM-186 FCM-058 FCM-069 FCM-068 FCM-070 FCM-070 FCM-166 FCM-152 FCM-159 FCM-167 FCM-068 FCM-153 FCM-170 FCM-155 FCM-170 FCM-171 FCM-136 FCM-158 FCM-071 FCM-071 FCM-187 FCM-134 FCM-154 FCM-072 FCM-067 FCM-067 FCM-060 FCM-164 FCM-059 FCM-149 FCM-060 FCM-135 FCM-151 FCM-058 FCM-172 FCM-185 FCM-168 FCM-059
CA10158-456EA 420.47 CAD
10158-886 CA10158-456 10158-920 10158-884 CA10158-454 10158-882 10158-880 CA10158-890 10065-994 10158-918 10158-916 10065-998 10065-996 CA10158-888 CA10158-886 10158-878 CA10158-920 CA10158-884 10066-062 10158-876 CA10158-882 10158-874 CA10158-880 CA10158-482 CA10158-480 CA10066-874 CA10066-876 10146-658 10146-666 10146-662 CA10158-918 10146-664 CA10158-916 10146-660 CA10158-878 CA10158-876 CA10158-478 CA10158-874 10066-010 CA10066-780 10066-012 CA10066-782 10066-014 CA10158-470 CA10066-860 10146-724 CA10066-858 10146-656 10146-650 10146-652 10146-690 CA10158-508 10146-692 CA10158-506 CA10158-504 CA10158-468 CA10158-466 CA10158-464 CA10158-462 CA10158-460 10146-718 10158-890 10066-006 10066-008 CA10066-776 CA10066-856 CA10066-778 10146-688 10146-720 10146-722 10158-888 CA10158-458
ACTERO™ Other Application Enrichment Media, FoodChek Systems
Media for Microbiology Prepared Media for Microbiology
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.