ACTERO™ Salmonella Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC. CA
Food and environmental samples require enrichment before pathogen testing enabling target pathogen present to grow and be detected in test samples
Using Actero™ Elite Enrichment Media significantly reduces all sample enrichment phase timelines regardless the type of "food pathogen testing system," such as lateral flow, ELISA and PCR platforms.
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.
The patented Actero™ Elite Enrichment Media are enhanced formulations that reduce the enrichment time required for accurate pathogen testing. Actero™ Salmonella Enrichement Media recieved the AOAC-PTM (041403) accreditation to be used as single-step recover and enrichment of Salmonella spp. from selected food and environmental samples.