ACTERO™ Coliform Enrichment Media, FoodChek Systems
Supplier: FOODCHEK SYSTEMS INC. CA
FCM-079 FCM-092 FCM-073 FCM-093 FCM-080 FCM-080 FCM-091 FCM-079 FCM-074 FCM-074 FCM-075 FCM-092 FCM-081 FCM-075 FCM-081 FCM-073 FCM-091 FCM-093
CA10066-660EA 454.81 CAD
10066-040 10066-064 10066-020 10066-066 CA10066-660 10066-036 CA10066-784 10066-038 10066-016 CA10066-662 10066-028 CA10066-786 10066-018 CA10066-788 CA10066-658 CA10066-804 CA10066-806 CA10066-808
ACTERO™ Coliform Enrichment Media, FoodChek Systems
Media for Microbiology Prepared Media for Microbiology
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Actero™ Universal Enrichment Media are established formulations that are used in standard protocols employed in testing labs today.