Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]

Supplier: Tonbo Biosciences
20-0059-T025 20-0059-T100
10140-994EA 126.4 CAD
10140-994 10140-996
Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Antibodies
The UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.

Type: Primary
Antigen: CD5
Clonality:
Clone: UCHT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR