KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Heavy Duty, Wide Mouth, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
92820G-5 92820G-10 92820G-20 92820G-25 92820G-50 92820G-100 92820G-200 92820G-250 92820G-500 92820G-1000 92820G-2000
89217-680CS 623.9 CAD
89217-680 89217-682 89217-684 89217-686 89217-688 89217-690 89217-692 89217-694 89217-696 89217-698 89217-700
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Heavy Duty, Wide Mouth, Kimble Chase
These KIMAX® flasks are permanently marked with an individual serial number and are supplied with a Certificate of Graduation Accuracy
Designed from ASTM Specification E288, Class A serialized requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Pipet access is easy with these wide-mouth volumetric flasks. Heavy, uniform walls provide superior strength, durability and safety. Large, permanent, easy-to-read markings allow for quick identification. Graduated to Class A volumetric tolerances with graduation ring blasted on the neck. Calibrated "to contain." Flasks feature flat interior bottoms for stirring with standard stir bars. Supplied with a marking spot and standard taper glass stopper.