Water USP, Ph. Eur., purified

Supplier: Ricca Chemical

R9190000-10F R9190000-20F R9190000-4A 9190-5HP 9190-1G R9190000-4F
RC9190-2.5EA 227.52 CAD
RC9190-2.5 RC9190-5 RC9190-1 RC91905HP RC9190-1G RCR9190000-4F
Water USP, Ph. Eur., purified
Water

This product is manufactured by a continuous process that has been validated under cGMP to produce high purity Water meeting, as a minimum, the specifications below at time of packaging.
This product has a low microbial count, but is not filled in a sterile area or into sterilized containers. Therefore, this water is not sterile and should not be used for injection or parenteral solution preparation.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Acidity or Alkalinity To Pass Test
Ammonia (NH₃) max 0.3 ppm
Ammonium (NH₄) max 0.2 ppm
Appearance Clear, colorless liquid
Calcium (Ca) max 0.5 ppm
Carbon Dioxide (CO₂) To Pass Test
Chloride (Cl) max 0.5 ppm
Conductivity at 25°C (Method: SQCP031) max 1 µS/cm
Endotoxins (Chromogenic determination) max 0.25 EU/mL
Heavy Metals (as Pb) max 0.1 ppm
Magnesium (Mg) max 0.5 ppm
Microbial Count (at time of manufacture) max 100 CFU/mL
Nitrate (NO₃) max 0.2 ppm
Organic Carbon (TOC) max 0.50 ppm
Oxidizable Substances (Permanganate Retention) To Pass Test
pH at 25°C (Method: SQCP027) 5.0-7.0
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Sulfate (SO₄) max 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR