Water ACS, reagent ASTM Type II
Supplier: Ricca Chemical
Synonyms:
Hydrogen oxide
9152-1
9152-32
9152-5
R9152000-10F
R9152000-55E
R9152000-500A
RC9152-1EA
74.26
CAD
RC9152-1
RC9152-32
RC9152-5
RCR9152000-10F
RCR9152000-55E
RCR9152000500A
Water ACS, reagent ASTM Type II
Water
Packaged in plastic. Not suitable for applications requiring extremely low levels of organics. This product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, the specifications below at the time of packaging. Although this water is not distilled, it has a chemical purity equal to or better than double distilled water.
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Density: 0.997 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 13,10098 |
Specification Test Results
Appearance | Colorless liquid |
Chloride (Cl) | max 5 ppb |
Conductivity at 25°C (Method: SQCP031) | max 1.0 µS/cm |
Heavy Metals (as Pb) | max 0.01 ppm |
Nitrate (NO₃) | max 0.4 ppm |
Organic Carbon (TOC) | max 50 ppb |
Oxidizable Substances (Permanganate Retention) | To Pass Test |
Phosphate (PO₄) | max 1.0 ppm |
Silicate (as SiO₂) | max 3 ppb |
Sodium (Na) | max 5 ppb |
Sulfate (SO₄) | max 1.0 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...